Imprimer
Cover glasses
Fournisseur: GLASWARENFABRIK KARL HECHT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
92101401190 92101501080 92101401120
HECH92101401190EA 0 EUR
HECH92101401190 HECH92101501080 HECH92101401120
Cover glasses
Lamelles couvre-objet
High spectral transmission, excellent flatness, optimum resistance to chemicals.
  • Free from colour distortion borosilicate glass D 263™ M
  • Excellent quality: Low levels of inclusions, striae, bubbles and streaks
Certifications: ISO 8255
Cdt: In tight-closing plastic boxes for protection against damp and climatic influences.