Imprimer
HPV type 16/18 probe, PATHO-GENE®
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ENZ32886 ENZ328866000
ENZOENZ32886EA 787 EUR
ENZOENZ32886 ENZOENZ328866000
HPV type 16/18 probe, PATHO-GENE®
Assays Immunological Assays
The PATHO-GENE® HPV type 16/18 probe is a mixture of biotin-labeled HPV 16 and HPV 18-specific probes in buffered formamide and hybridisation enhancers.
The ENZO PATHO-GENE® HPV type 16/18 probe employs a mixture of the HPV 16 and HPV 18-specific probes that detect HPV types that have been associated with cervical intraepithelial neoplasia (CIN) and carcinoma in situ (CIS). The probe mixture is denatured and hybridised to the tissue section fixed and pretreated on the microscope slide. Specific hybridisation between the HPV DNA probe and DNA in the specimen is determined by the detection of biotin.