Imprimer
Restriction enzymes, HindIII, 10 U/μl, Fermentas
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ER0501 ER0502 ER0505
ER0501EA 39.94 EUR
ER0501 ER0502 ER0505
Restriction enzymes, HindIII, 10 U/μl, Fermentas
Nucleic Acid Reagents Restriction Enzymes
HindIII restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions.
  • Convenient color-coded Five Buffer System
  • It includes universal Tango buffer for double-digestions
  • Reaction buffers contain premixed BSA
  • Wide selection of restriction endonuclease specificities
Recognises A^AGCTT sites and cuts best at 37 °C.