Fluorescent dyes, M series, streptavidin conjugate
Marques : AbnovaTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Fluorescent dye, M series, products are designed to be maximally excited by one of the major light sources equipped in flow cytometers. In comparison with phycobiliprotein tandems, Fluorescent dye, M series, possesses better photostability, higher conjugation yields, and little pH sensitivity making it an ideal choice in fluorescence imaging applications.
Recommended usage: Labeling via streptavidin for fluorescence imaging and other fluorescence-based biochemical analysis. For most fluorescent streptavidin applications, a concentration of 1 to 20 g/ml is sufficient. The optimal working dilution should be determined by the end user.