Imprimer
Reagents for AP3900
Marques : Hach
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
APC114 APC342 APC341 APC138 APC238 APC303 APC338 APC339 APC340 APC348 APC350 APC304 APC349 APC314
HACHAPC114EA 586 EUR
HACHAPC114 HACHAPC342 HACHAPC341 HACHAPC138 HACHAPC238 HACHAPC303 HACHAPC338 HACHAPC339 HACHAPC340 HACHAPC348 HACHAPC350 HACHAPC304 HACHAPC349 HACHAPC314
Reagents for AP3900
Kits de tests environnementaux
Tests for all water analysis applications. The cuvette is a closed sytem which is safe for the user and the environment. Suitable for the laboratory robot.
  • Accurate and precise measured values
  • Fast and simple procedures
  • Convenient and error-free dosing of the reagents
  • Pre-dosed reagents for maximum safety
Differentiation between tests and measuring ranges by means of colour-coding. Also barcode labelled for automatic recognition in the photometer.