Imprimer
Hubbell plugout for industrial plug connections
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
065968 065695
BRDY065968EA 76.3 EUR
BRDY065968 BRDY065695
Hubbell plugout for industrial plug connections
Cadenas de sécurité
These Hubbell plugouts effectively secure all larger industrial plug connections.
  • Two or four padlocks can be applied
Max. shackle diameter: 7,00 mm
Min. shackle clearance: 20,00 mm