Imprimer
Bags, zip seal, anti-UV, writing area, Minigrip®
Fournisseur: Flexico
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ITM025704 ITM025701 ITM025702
129-2360EA 474 EUR
129-2360 129-3876 129-3877
Bags, zip seal, anti-UV, writing area, Minigrip®
Sachets Sachets d'échantillonnage
PE bags, thickness 80 µm.
  • UV protection
  • 3 white write-on panels
  • Resealable, reusable, 100% recyclable
Made of polyethylene (LDPE). UV protection provides barrier properties for the content of the bag (wavelengths between 280 and 350 nm) and can withstand an irradiation energy of 200 Kly. Zip seal designed for easy opening and high speed packing. The Minigrip® bag provides an excellent mechanical resistance and high temperature resistance (from −40°C up to +60°C). NOT suitable for food contact.