Imprimer
Surface cleaner for smart phones, PhoneSoap
Fournisseur: Dino-Lite / IDCP B.V.
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PS500-3B PS500-3W
IDCPPS500-3BEA 0 EUR
IDCPPS500-3B IDCPPS500-3W
Surface cleaner for smart phones, PhoneSoap
Produits nettoyants Détergents et Nettoyants de Surfaces
UV sanitisers for smartphones that kill all the bacteria growing on your device.
  • UV Sanitiser
  • Can charge your device while it sanitises
  • Sanitising in less than 15 minutes
  • Blue indicator light when sanitising is in progress
Powerful UV-C lights (254 nm) and a reflective inner surface allow the light to surround your device
PhoneSoap 2.0 (for smart phones): Inner Dimensions (Sanitizing Space L×W×H) 173×95×20 mm
PhoneSoap 2.0 has a standard 5V USB port accessible on the back of the device next to the unit’s power port. You will use the cable provided with your phone to charge.
PhoneSoap XL (for tablets): Inner Dimensions (Sanitizing Space L×W×H) 314×91×247 mm
PhoneSoap XL has a 2,4A USB port accessible on the back of the device next to the unit’s power port. You will use the cable provided with your tablet to charge.
Informations de livraison: PhoneSoap 2.0, power plug with USB to Micro USB cord to power the device.
PhoneSoap XL with a wall port with a mirco USB to USB cord used to plug into the wall port to power the device.