Imprimer
Trays, SteriWare®
Fournisseur: Sampling Systems
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8081P-3000S 8081P-3001
SAMP8081P-3000SEA 633 EUR
SAMP8081P-3000S SAMP8081P-3001
Trays, SteriWare®
Plateaux/Récipients/Pots Plateaux et récipients de laboratoire
Trays made of PP, translucent.
  • Assembled and packed in a cleanroom
  • Individually bagged
  • FDA & EU 10/2011 conforming materials
  • BSE/TSE free
  • Full batch traceability
  • Available sterile (gamma irradiated)
The disposable tray is ideal for hygienic applications within hospitals and pharmaceutical factories.