Imprimer
Rocker cell culture bags, Corning®
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
91-200-92 91-200-78 91-200-79 91-200-80 91-200-81
MDTC91-200-92EA 510 EUR
MDTC91-200-92 MDTC91-200-78 MDTC91-200-79 MDTC91-200-80 MDTC91-200-81
Rocker cell culture bags, Corning®
Bags Media Bags
Rocker cell culture bags are designed for use with commercial rocking motion bioreactor chambers, and feature irradiated ethylene vinyl acetate (EVA)/low-density polyethylene (ULDPE) 9101 film and stability bars on each side. These sterile, single-use cell culture bags are ideal for applications from basic research to large-scale biopharmaceutical manufacturing.
  • Provides non-invasive mixing of the cell culture/media with efficient gas transfer
  • Eliminates the risk of cross-contamination
  • Used for basic research to large scale cell culture applications
  • Available in choice of sizes: 2, 10, 20, 22 and 50 L
The rocking motion of the bioreactor platform creates effective, non invasive mixing of the cell culture/media with efficient gas transfer. Rocking-motion prevents cells from settling at the bottom of the rocker cell culture bag while generating large amounts of recombinant protein in both mammalian and insect cell lines.
Port configurations:
Port 1 (all bag sizes)
Check valve with ¹/₄" barb
Pall 8004022 filter
2" length ³/₁₆" Int.Ø. × ³/₈" Ext.Ø. PCMGS tube
Port 2 (all bag sizes)
Pall 8004022 filter
2" length ³/₁₆" Int.Ø. × ³/₈" Ext.Ø. PCMGS tube
Port 3 (2 L, 10 L, 20 L, 50 L bags)
Female MPC connector with ¹/₄" barb
Male MPC plug
40" length ¹/₄" Int.Ø. × ⁷/₁₆" Ext.Ø. chemically resistant, heat sealable flexible tube
Port 3 (22 L bag only)
Female MPC connector with ¹/₄" barb
Male MPC plug
48" length ¹/₄" Int.Ø. × ⁷/₁₆" Ext.Ø. chemically resistant, heat sealable flexible tube
Port 4 (2 L, 10 L, 20 L, 50 L bags)
Female Luer with ¹/₄" barb
Male Luer plug
40" length ¹/₈" Int.Ø. × ¹/₄" Ext.Ø. chemically resistant, heat sealable flexible tube
Port 4 (22 L bag only)
Female Luer with ¹/₄" barb
Male Luer plug
48" length ¹/₈" Int.Ø. × ¹/₄" Ext.Ø. chemically resistant, heat sealable flexible tube
Port 5 (all bag sizes)
Female Luer with ¹/₄" barb
Swabbable valve
Male Luer plug without stem
Informations de commande: Optional accessories and tubing sets are available.