Imprimer
Rack for sedimentation plates
Fournisseur: MERCK LIFE SCIENCE GMBH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HEIP1465190001EA 157 EUR
HEIP1465190001
Rack for sedimentation plates
Portoirs Distributeurs de boîtes de Petri
High-quality stainless steel sedimentation plate rack suitable for isolators and cleanroom applications (resistant against H₂O₂ and disinfectants).
  • Locking device aids safe, convenient handling
  • Autoclavable