Imprimer
Cell scrapers and cell lifters
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3008 3010 3011
734-1526EA 305 EUR
734-1526 734-1527 734-1528
Cell scrapers and cell lifters
Cell Scrapers
Useful for the manual harvesting of cells, scrapers are designed for use in flasks and cell lifters for use in harvesting cells (especially stem cells) in dishes.
  • Blade design minimises cell damage and ensures even contact with the growth surface
  • Individually wrapped
  • Sterilised by gamma radiation
  • Certified non-pyrogenic