Imprimer
Cleanroom wipes, VeraClean®
Fournisseur: PGI NONWOVEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
74607 74609 74608
115-2502EA 564 EUR
115-2502 PGIN74609 PGIN74608
Cleanroom wipes, VeraClean®
Lingettes Lingettes pour salles propres Lingettes pour salles propres, sèches
These wipes are ideal for use in critical production processes, such as semi-conductors, pharmaceuticals, biotechnology and all other environments where contamination is a possibility.
  • Low levels of ions and organic extractables
  • Highly absorbent for effective cleaning
  • Low linting
Cdt: Supplied in durable double sealed packaging.