Imprimer
Agarose, NuSieve® 3:1
Fournisseur: LONZA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
50094 50090 50091
733-1209EA 4070 EUR
733-1209 733-1207 733-1208
Agarose, NuSieve® 3:1
Gels d'électrophorèse Agarose et acrylamide
NuSieve® 3:1 is a high strength agarose ideal for analytical electrophoresis of small DNA/RNA fragments and in-gel PCR/transformation/ligation.
  • High gel strength makes gels easy to handle, enhancing the convenience of gel processing and blotting
  • Performance tested to ensure fine resolution of DNA fragments up to 1000 bp
  • DNAse- and RNAse-free