Imprimer
Agarose, SeaKem® HEEO
Fournisseur: LONZA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
50030 50031
733-1557EA 1030 EUR
733-1557 733-1558
Agarose, SeaKem® HEEO
Gels d'électrophorèse Agarose et acrylamide
SeaKem® HEEO Agarose is a very high EEO agarose that is useful in applications requiring significant cathodal migration, such as IEP of IgG and IgM. It can also be used to blend with a lower EEO agarose to achieve a specific EEO value.