Imprimer
Agarose, SeaKem® HE
Fournisseur: LONZA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
50020 50021
733-1555EA 815 EUR
733-1555 733-1556
Agarose, SeaKem® HE
Gels d'électrophorèse Agarose et acrylamide
SeaKem® HE agarose is a high EEO agarose that provides enhanced resolution in immunoelectrophoresis (IEP), crossed-IEP, counterimmunoelectrophoresis (CIEP), and serum protein electrophoresis.