Imprimer
Lamelles couvre-objets, Menzel-Gläser
Fournisseur: Epredia
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BB022022A153 BB022022AC53 BB020020A153 BB020020AC53 BC038038A140 BC050055A140 BB015015A123 BB015015A1D3 BB024024A153 BB024024AC23 BB018018AC20 BB020020A123 BB020020AC23 BB022022A123 BB022022A2D3 BB024024A123 BC025025A113 BB024024AC53 BC032032A140 BC048048A140 BB018018A2 BB015015A2D3 BC044044A140
631-1336EA 43.9 EUR
631-1336 631-0851 631-1334 630-2101 MENZBC038038A140 MENZBC050055A140 MENZBB015015A123 631-0710 631-1337 630-2105 MENZBB018018AC20 630-2888 630-2886 630-2882 630-2881 630-2104 MENZBC025025A113 630-2106 630-0953 MENZBC048048A140 MENZBB018018A2 MENZBB015015A2D3 MENZBC044044A140
Lamelles couvre-objets, Menzel-Gläser
Lamelles couvre-objet
Pre-washed pure white D 263 M borosilicate glass of hydrolytic class 1 cover slips.
  • High spectral transmission, uniform quality of thickness
  • No bubbles or flaws guarantees high optical homogeneity
  • Highly transparent and achromatic: Practically free of any colour distortion
Suitable for in vitro diagnostic (IVD) applications in medicine, biology and research examinations.
Certifications: DIN ISO 719 ; DIN ISO 12 116 ; DIN ISO 695.