Imprimer
Finned plugs for tubes
Fournisseur: KARTELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
88305-00 8830504 8830512 8830510 8830506 88309-00 88309-04 88309-10 88309-06 88309-12
212-0540EA 51.1 EUR
212-0540 212-0541 212-0544 212-0543 212-0542 212-0545 212-0546 212-0548 212-0547 212-0549
Finned plugs for tubes
Bouchons Bouchons en plastique
Finned plugs for disposable tubes 11 to 13 mm or 15 to 17 mm in diameter. The finned plug has been carefully designed. Available colours: Neutral (00), blue (04), red (10), green (12) and yellow (06).