Imprimer
AlphaTec® 37-510/37-646 General Purpose Gloves
Fournisseur: Ansell
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
37-646/12 37-646/9
ANSE37-646/12EA 0 EUR
ANSE37-646/12 ANSE37-646/9
AlphaTec® 37-510/37-646 General Purpose Gloves
Gants
Unlined nitrile gloves especially designed for use in the food industry.
  • High performance nitrile compound for enhanced flexibility and dexterity, even in cold environments
  • Ansell UltraGrip™ technology: thanks to the raised diamond grip pattern, these gloves offer unmatched grip in wet and fatty conditions
  • Latex free formulation: no risk of Type I allergies
  • Unflocked: to prevent the risk of food contamination
  • Offer superior mechanical properties over natural rubber gloves
Ideal for use in food preparation, processing and handling, meat and poultry processing, fish processing and cleaning.
Certifications: EN 388:2016 (2001X), EN ISO 374-1 Type B (JKL), EN ISO 374-5, EN 421; Cat. III.
Food contact approved.