Imprimer
General purpose gloves, HyFlex® 48-102
Fournisseur: Ansell
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
48-102/7 48-102/8 48-102/9 48-102/10 48-102/11
ANSE48-102/7EA 34.3 EUR
ANSE48-102/7 ANSE48-102/8 ANSE48-102/9 ANSE48-102/10 ANSE48-102/11
General purpose gloves, HyFlex® 48-102
Gants
Seamless knitted polyamide gloves with PU coating on palm and fingertips.
  • Textile liner made of high-tenacity polyamid yarn
  • Fine and stretchable stitches for an excellent comfort
  • Stretchable palm fit PU coating
  • Good abrasion resistance
  • Low linting
Ideal for delicate and precise operations, finishing works, general handling and packaging.
Certifications: EN 388:2016 (4121A); Cat. II.