Dideoxyuridine triphosphates (ddUTPs)
Marques : ENZO LIFE SCIENCESTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
These ddUTPs are used for 3’-end labeling of oligodeoxynucleotides using terminal transferase. ddUTP also serves as a substrate for E. coli DNA Polymerase I (holoenzyme and Klenow fragment), T4 and T7 DNA polymerases, Taq DNA polymerase and reverse transcriptase.
- Purity ≥93% measured by HPLC
- Purified by ion-exchange chromatography
Oligonucleotide probes labeled with (6) TAMRA-12-ddUTP and (6) ROX-12-ddUTP can be detected by direct fluorescence detection (orange fluorescence and red fluoroscence respectively) or by methods using horseradish peroxidase- or alkaline phosphatase-linked anti-fluorescein antibodies.