Imprimer
Microsurgical blades, Feather®
Fournisseur: PFM Medical GmbH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
200600541 200600560 200600561 200600511 200600530
FEAT200600541EA 0 EUR
FEAT200600541 FEAT200600560 FEAT200600561 FEAT200600511 FEAT200600530
Microsurgical blades, Feather®
Rasoirs et lames
These stainless steel precision blades are part of a microsurgical system of inter-changeable blades and handles for flexible and efficient work.
  • Made of high-quality stainless steel using high-precision grinding technology
  • Optimised grinding technology and carefully selected materials produce consistent, ultra-sharp cutting edges
  • Consistent incision size minimises the risk of tissue injury during incision
  • Blades are gammasterilised and can be used immediately after opening the package
  • Protective guard (blade guide) prevents any damage to the underlying tissue during incisions (only valid for some blades)
  • Handles are made from titanium alloy
Informations de commande: Blades and handles need to be ordered separately.