Imprimer
Labels for BBP3x printers, type B-581
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
118009 118008 118005 118003 118004 118006 118007 118010
BRDY118009EA 198 EUR
BRDY118009 BRDY118008 BRDY118005 BRDY118003 BRDY118004 BRDY118006 BRDY118007 BRDY118010
Labels for BBP3x printers, type B-581
Étiquettes
These repositionable vinyl B-581 labeling tapes with removable acrylic are ideal for temporary, removable, repostionable labeling applications.