Imprimer

Anti-NAPSA Mouse Monoclonal Antibody (CF594) [clone: NAPSA/1239]

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BNC941239-500 BNC941239-100
BNC941239-500EA 560 EUR
BNC941239-500 BNC941239-100
Anti-NAPSA Mouse Monoclonal Antibody (CF594) [clone: NAPSA/1239]
Anticorps
Napsin is a pepsin-like aspartic proteinase connected with maturation of surfactant protein B.There are two closely related napsins, napsin A and napsin B. Napsin A is expressed as a single chain protein. Immunohistochemical studies revealed high expression levels of napsin A in human lung and kidney but low expression in spleen. Napsin A is expressed in type II pneumocytes and in adenocarcinomas of lung. The high specificity expression of napsin A in adenocarcinomas of lung is useful to distinguish primary lung adenocarcinomas from adenocarcinomas of other organs.

Type: Primary
Antigen: NAPSA
Clonality: Monoclonal
Clone: NAPSA/1239
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR