Imprimer

Anti-UACA Mouse Monoclonal Antibody (CF647) [clone: UACA/1222]

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BNC471222-500 BNC471222-100
BNC471222-500EA 560 EUR
BNC471222-500 BNC471222-100
Anti-UACA Mouse Monoclonal Antibody (CF647) [clone: UACA/1222]
Anticorps
UACA (Uveal Autoantigen with Coiled-coil domains and Ankyrin repeats) is a 1,416 amino acid nuclear membrane protein. It was originally identified as an autoantigen in patients with panuveitis, a characteristic of Vogt-Koyanagi-Harada disease, and in patients with Graves' disease. UACA was also later identified as Nucling, an mRNA differentially expressed in F9 embryonal carcinoma cells during cardiac muscle differentiation. UACA appears to function as a pro-apoptotic protein that recruits the apaf-1-pro-caspase-9 complex for the induction of apoptosis to mediate the cell-death pathway.

Type: Primary
Antigen: UACA
Clonality: Monoclonal
Clone: UACA/1222
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR