Imprimer

Anti-MCAM Mouse Monoclonal Antibody (CF488A) [clone: MUC18/1130]

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BNC881130-500 BNC881130-100
BNC881130-500EA 560 EUR
BNC881130-500 BNC881130-100
Anti-MCAM Mouse Monoclonal Antibody (CF488A) [clone: MUC18/1130]
Anticorps
The human Mel-CAM gene maps to chromosome 11q23 and encodes a trans-membrane glycoprotein, also designated MCAM, MUC 18 or CD146, that belongs to the immunoglobulin superfamily and functions as a Ca2 -independent cell adhesion molecule. Mel-CAM expression is restricted to advanced primary and metastatic melanomas and to cell lines of the neuroectodermal lineage, but not normal melanocytes. Mel-CAM is found on 80% of advanced primary human melanomas and correlates well with development of metastatic disease.

Type: Primary
Antigen: MCAM
Clonality: Monoclonal
Clone: MUC18/1130
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR