Imprimer

Anti-Human Papillomavirus 16 Mouse Monoclonal Antibody (CF568) [clone: HPV16/1058]

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BNC681058-500 BNC681058-100
BNC681058-500EA 560 EUR
BNC681058-500 BNC681058-100
Anti-Human Papillomavirus 16 Mouse Monoclonal Antibody (CF568) [clone: HPV16/1058]
Anticorps
Reacts with a protein of 57 kDa, identified as the L1 protein of human papilloma virus type 16 (HPV-16). It is the major capsid protein of HPV-16. Infection with specific types of HPV has been associated with an increased risk of developing cervical neoplasia. HPV types 6 and 11 have been associated with relatively benign diseases such as genital warts but types 16 and 18 are strongly associated with cervical, vaginal, and vulvar malignancies. The antibody reacts very strongly with formalin-fixed, paraffin-embedded tissues containing HPV-16 or -33; very weak reactions were occasionally observed with biopsy specimens or smears containing HPV-6 or HPV-11. It cross-reacts with HPV37.

Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: HPV16/1058
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: HPV
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR