Reacts with a monomorphic determinant of human major histocompatibility (MHC) class I antigens (HLA-A, B and C). Human MHC class I antigens are expressed constitutively on all nucleated cells lymphocytes such as lymphocytes, thymocytes, granulocytes, and bone marrow cells and are absent on erythrocytes. MHC class I antigens play a role in class I MHC-associated antigen presentation, inhibition of NK cell cytotoxicity, tumor surveillance, and tissue allotransplantation.
Type: Primary
Antigen: HLA-A; HLA-B; HLA-C
Clonality: Monoclonal
Clone: 246-B8.E7
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human, Mouse, Non-human primates