Imprimer

Anti-Ep-CAM Mouse Monoclonal Antibody [clone: EGP40/826 EGP40/837 EGP40/1110 EGP40/1120]

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BNUB0969-500 BNUB0969-100
BNUB0969-500EA 511 EUR
BNUB0969-500 BNUB0969-100
Anti-Ep-CAM Mouse Monoclonal Antibody [clone: EGP40/826 EGP40/837 EGP40/1110 EGP40/1120]
Anticorps
This is a cocktail of four highly specific monoclonal antibodies (EGP40/826, EGP40/837, EGP40/1110, EGP40/1120) that recognize extracellular as well as intracellular domains of the epithelial cellular adhesion molecule (Ep-CAM). It is a 40-43 kDa transmembrane epithelial glycoprotein, identified as epithelial specific antigen (ESA), or Ep-CAM. Ep-CAM is expressed on baso-lateral cell surface in most simple epithelia and a vast majority of carcinomas. This epithelial antigen plays an important role as a tumor-cell marker in lymph nodes from patients with esophageal carcinoma otherwise classified as node-negative. Epithelial antigen has also been suggested as a discriminator between basal cell and baso-squamous carcinomas, and squamous cell carcinoma of the skin.

  • Purified

Type: Primary
Antigen: Ep-CAM
Clonality: Monoclonal
Clone: EGP40/826 EGP40/837 EGP40/1110 EGP40/1120
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR