Imprimer
Anti-KRT7 Mouse Monoclonal Antibody (CF488A) [clone: KRT7/903]
Fournisseur: Biotium
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BNC880903-500 BNC880903-100
BNC880903-500EA 560 EUR
BNC880903-500 BNC880903-100
Anti-KRT7 Mouse Monoclonal Antibody (CF488A) [clone: KRT7/903]
Anticorps
Cytokeratin 7 is a basic cytokeratin, which is found in most glandular and transitional epithelia but not in the stratified squamous epithelia. Keratin 7 is expressed in the epithelial cells of ovary, lung, and breast but not of colon, prostate, or gastrointestinal tract. Antibody to cytokeratin is useful in distinguishing ovarian carcinomas (keratin 7 ) from colon carcinomas (keratin 7-).
Type: Primary
Antigen: KRT7
Clonality: Monoclonal
Clone: KRT7/903
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human