Imprimer

Anti-AFP Mouse Monoclonal Antibody [clone: C2 C3 MBS-12]

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BNUB0810-500 BNUB0810-100
BNUB0810-500EA 511 EUR
BNUB0810-500 BNUB0810-100
Anti-AFP Mouse Monoclonal Antibody [clone: C2 C3 MBS-12]
Anticorps
This antibody recognizes an oncofetal glycoprotein with a single chain of 70 kDa, which is identified as alpha fetoprotein (AFP). This MAb is highly specific to AFP and shows no cross-reaction with other oncofetal antigens or serum albumin. AFP is normally synthesized in the liver, intestinal tract, and yolk sac of the fetus. Antibody to AFP has been shown to be useful in detecting hepatocellular carcinomas (HCC) and germ cell neoplasms, especially yolk sac tumors.

  • Purified

Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone: C2 C3 MBS-12
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR