Imprimer

Anti-TGFA Mouse Monoclonal Antibody (CF568) [clone: TG86]

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BNC680786-500 BNC680786-100
BNC680786-500EA 560 EUR
BNC680786-500 BNC680786-100
Anti-TGFA Mouse Monoclonal Antibody (CF568) [clone: TG86]
Anticorps
This antibody reacts with the TGF alpha and shows no cross-reaction with EGF and the neuropeptide synenkephalin. The staining with this MAb is completely blocked by the peptide used for raising this antibody. TGF alpha (aa50) is a growth factor with 33% homology to EGF, binds to EGFR, activates tyrosine phosphorylation of the receptor, and stimulates cell proliferation. It plays a role in tumor initiation by inducing the reversible transformed phenotype.

Type: Primary
Antigen: TGFA
Clonality: Monoclonal
Clone: TG86
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rabbit, Zebrafish
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR