Imprimer
Anti-CD68 Mouse Monoclonal Antibody (CF594) [clone: C68/684]
Fournisseur: Biotium
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BNC940684-500 BNC940684-100
BNC940684-500EA 560 EUR
BNC940684-500 BNC940684-100
Anti-CD68 Mouse Monoclonal Antibody (CF594) [clone: C68/684]
Anticorps
This antibody recognizes a glycoprotein of 110 kDa, which is identified as CD68. It is important for identifying macrophages in tissue sections. It stains macrophages in a wide variety of human tissues, including Kupffer cells and macrophages in the red pulp of the spleen, in lamina propria of the gut, in lung alveoli, and in bone marrow. It reacts with myeloid precursors and peripheral blood granulocytes. It also reacts with plasmacytoid T cells, which are supposed to be of monocyte/macrophage origin. It shows strong granular cytoplasmic staining of chronic and acute myeloid leukemia andEn savoir plus