Imprimer

Anti-CSF2 Mouse Monoclonal Antibody (CF594) [clone: BVD2-21C11]

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BNC940662-500 BNC940662-100
BNC940662-500EA 560 EUR
BNC940662-500 BNC940662-100
Anti-CSF2 Mouse Monoclonal Antibody (CF594) [clone: BVD2-21C11]
Anticorps
Granulocyte/macrophage - Colony-stimulating factor (GM-CSF) is a hematopoietic factor that is produced by activated T-cells, B-cells, mast cells, macrophages, fibroblasts, and endothelial cells. In addition to supporting colony formation of granulocyte/macrophage progenitors, GM-CSF is a growth factor for erythroid, megakaryocyte, and eosinophil progenitors.

Type: Primary
Antigen: CSF2
Clonality: Monoclonal
Clone: Clone BVD2-21C11
Conjugation: CF594
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Cynomolgus monkey, Human, Rhesus monkey
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR