Imprimer

Anti-MYC Mouse Monoclonal Antibody (CF647) [clone: 9E10.]

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BNC470596-500 BNC470596-100
BNC470596-500EA 560 EUR
BNC470596-500 BNC470596-100
Anti-MYC Mouse Monoclonal Antibody (CF647) [clone: 9E10.]
Anticorps
This antibody recognizes a transcription factor of 64-67 kDa, identified as c-myc. Its epitope spans between aa 410-419 (EQKLISEEDL) which is a specific portion of an alpha helical region of human c-myc protein. This MAb shows no cross-reaction with v-myc. c-myc is involved in the control of cell proliferation and differentiation and is amplified and/or overexpressed in a variety of tumors. Over-expression of c-myc protein occurs frequently in luminal cells of prostate intraepithelial neoplasia as well as in most primary carcinomas and metastatic disease.

Type: Primary
Antigen: MYC
Clonality: Monoclonal
Clone: 9E10.
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR