Imprimer
Anti-PGR Mouse Monoclonal Antibody (CF640R) [clone: PR501]
Fournisseur: Biotium
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BNC400501-500 BNC400501-100
BNC400501-500EA 560 EUR
BNC400501-500 BNC400501-100
Anti-PGR Mouse Monoclonal Antibody (CF640R) [clone: PR501]
Anticorps
This MAb is specific to progesterone receptor and shows minimal cross-reaction with other members of the family. Progesterone receptor is expressed as two major isoforms, PR-A (81 kDa) and PR-B (116 kDa). Expression of PgR has been suggested to reflect a intact estrogen regulatory machinery and therefore, predict better clinical response to endocrine therapy than ER alone.
Type: Primary
Antigen: PGR
Clonality: Monoclonal
Clone: PR501
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human