Imprimer

Anti-PGR Mouse Monoclonal Antibody (CF640R) [clone: PR500]

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BNC400500-500 BNC400500-100
BNC400500-500EA 560 EUR
BNC400500-500 BNC400500-100
Anti-PGR Mouse Monoclonal Antibody (CF640R) [clone: PR500]
Anticorps
This MAb is specific to progesterone receptor and shows minimal cross-reaction with other members of the family. Progesterone receptor is expressed as two major isoforms, PR-A (81 kDa) and PR-B (116 kDa). Expression of PgR has been suggested to reflect a intact estrogen regulatory machinery and therefore, predict better clinical response to endocrine therapy than ER alone.

Type: Primary
Antigen: PGR
Clonality: Monoclonal
Clone: PR500
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR