Imprimer

Anti-ESR1 Mouse Monoclonal Antibody (CF488A) [clone: ESR1/420]

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BNC880420-500 BNC880420-100
BTIUBNC880420-500EA 560 EUR
BTIUBNC880420-500 BTIUBNC880420-100
Anti-ESR1 Mouse Monoclonal Antibody (CF488A) [clone: ESR1/420]
Anticorps
This MAb is specific to ER alpha and shows minimal cross-reaction with other members of the family. Epitope of this MAb is mapped between aa300-550. ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.

Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone: ESR1/420
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR