Imprimer
Anti-ESR1 Mouse Monoclonal Antibody (CF405S) [clone: ESR1/420]
Fournisseur: Biotium
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BNC040420-500 BNC040420-100
BTIUBNC040420-500EA 560 EUR
BTIUBNC040420-500 BTIUBNC040420-100
Anti-ESR1 Mouse Monoclonal Antibody (CF405S) [clone: ESR1/420]
Anticorps
This MAb is specific to ER alpha and shows minimal cross-reaction with other members of the family. Epitope of this MAb is mapped between aa300-550. ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.
Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone: ESR1/420
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human