Imprimer

Anti-Ep-CAM Mouse Monoclonal Antibody (Biotin) [clone: 323/A3]

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BNCB0396-500 BNCB0396-100
BTIUBNCB0396-500EA 560 EUR
BTIUBNCB0396-500 BTIUBNCB0396-100
Anti-Ep-CAM Mouse Monoclonal Antibody (Biotin) [clone: 323/A3]
Anticorps
EGP40 is a 40-43 kDa transmembrane epithelial glycoprotein, also identified as epithelial specific antigen (ESA), or epithelial cellular adhesion molecule (Ep-CAM). It is expressed on baso-lateral cell surface in most simple epithelia and a vast majority of carcinomas. This antibody has been used to distinguish adenocarcinoma from pleural mesothelioma and hepatocellular carcinoma. This antibody is also useful in distinguishing serous carcinomas of the ovary from mesothelioma.

Type: Primary
Antigen: Ep-CAM
Clonality: Monoclonal
Clone: 323/A3
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR