Imprimer

Anti-KRT77 & KRT76 Mouse Monoclonal Antibody (Purified) [clone: AE-1 / AE-3]

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BNUB0371-500 BNUB0371-100
BNUB0371-500EA 511 EUR
BNUB0371-500 BNUB0371-100
Anti-KRT77 & KRT76 Mouse Monoclonal Antibody (Purified) [clone: AE-1 / AE-3]
Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI 6.0) subfamilies. This antibody cocktail recognizes acidic (Type I or LMW) and basic (Type II or HMW) cytokeratins, which 67 kDa (CK1); 64 kDa (CK3); 59 kDa (CK4); 58 kDa (CK5); 56 kDa (CK6); 52 kDa (CK8); 56.5 kDa (CK10); 50 kDa (CK14); 50 kDa (CK15); 48 kDa (CK16); 40 kDa (CK19). Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis. AE-1/AE-3 is a broad spectrum anti pan-cytokeratin antibody cocktail, which differentiates epithelial tumors from non-epithelial tumors e.g. squamous vs. adenocarcinoma of the lung, liver carcinoma, breast cancer, and esophageal cancer. It has been used to characterize the source of various neoplasms and to study the distribution of cytokeratin containing cells in epithelia during normal development and during the development of epithelial neoplasms. This antibody stains cytokeratins present in normal and abnormal human tissues and has shown high sensitivity in the recognition of epithelial cells and carcinomas.

Type: Primary
Antigen: KRT77 & KRT76
Clonality: Monoclonal
Clone: AE-1 / AE-3
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Chicken, Cow, Dog, Human, Monkey, Mouse, Rabbit, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR