Imprimer

Anti-CD3E Mouse Monoclonal Antibody (CF647) [clone: RIV9]

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BNC470322-500 BNC470322-100
BNC470322-500EA 560 EUR
BNC470322-500 BNC470322-100
Anti-CD3E Mouse Monoclonal Antibody (CF647) [clone: RIV9]
Anticorps
Reacts with five invariable CD3 chains (designated as and ) with molecular weight ranging from 16-28 kDa. It is found on all mature T-lymphocytes, NK, T-cells, and some thymocytes. CD3, also known as T3, is a member of the immunoglobulin superfamily that plays a role in antigen recognition, signal transduction, and T cell activation. It is expressed, typically at high levels, on peripheral T cells and majority of T cell neoplasms. Thymocytes express CD3 at different level on the cell surface in the course of differentiation and, in cortical thymus, CD3 is predominantly Intracytoplasmic. This MAb is particularly useful for induction of T cell activation in vitro and in vivo due to its unusual IgG3 isotype.

Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: RIV9
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG3 kappa
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR