Imprimer

Anti-FN1 Mouse Monoclonal Antibody (CF594) [clone: HFN7.1]

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BNC940270-500 BNC940270-100
BNC940270-500EA 560 EUR
BNC940270-500 BNC940270-100
Anti-FN1 Mouse Monoclonal Antibody (CF594) [clone: HFN7.1]
Anticorps
Fibronectin is a dimeric glycoprotein of 440 kDa, which is present in cells, extracellular matrix, and blood. It possesses at least four binding sites for collagen, glycosaminoglycans, transglutaminase, and a cell surface receptor. Fibronectin is involved in cell adhesion, tissue organization, and wound healing. This MAb is directed against the peptide core and reacts with both the plasma and cellular forms of fibronectin. It blocks the fibronectin-medicated cell attachment not by disrupting the collagen-fibronectin interaction, but by interfering with the attachment of fibronectin to its receptor on the cell surface.

Type: Primary
Antigen: FN1
Clonality: Monoclonal
Clone: HFN7.1
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR