Imprimer

Anti-ICAM3 Mouse Monoclonal Antibody (Purified) [clone: 186-2G9]

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BNUB0178-500 BNUB0178-100
BNUB0178-500EA 511 EUR
BNUB0178-500 BNUB0178-100
Anti-ICAM3 Mouse Monoclonal Antibody (Purified) [clone: 186-2G9]
Anticorps
Recognizes the D1 domain of an N-glycosylated glycoprotein of 120 kDa with intra-chain disulfide bonds, identified as CD50 or ICAM-3 (Leucocyte Workshop VI). CD50 is the major ligand for LFA-1 (CD11a/CD18) and may have signaling role to increase adhesion. It is expressed on thymocytes and T lymphocytes and is resistant to treatment with phosphatidylinositol (PI) phospholipase C. This MAb is excellent for staining of formalin/paraffin tissues.

Type: Primary
Antigen: ICAM3
Clonality: Monoclonal
Clone: 186-2G9
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR