Imprimer

Anti-ICAM3 Mouse Monoclonal Antibody (CF568) [clone: 101-1D2]

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BNC680177-500 BNC680177-100
BNC680177-500EA 560 EUR
BNC680177-500 BNC680177-100
Anti-ICAM3 Mouse Monoclonal Antibody (CF568) [clone: 101-1D2]
Anticorps
Recognizes an N-glycosylated glycoprotein of 120 kDa with intra-chain disulfide bonds, identified as CD50 or ICAM-3 (WS: IV & V). Its epitope localizes in the D2 extracellular domain and is resistant to neuraminidase and proteases. CD50 is the major ligand for LFA-1 (CD11a/CD18) and may have signalling role to increase adhesion. It is expressed on thymocytes and T lymphocytes and is resistant to treatment with phosphatidylinositol (PI) phospholipase C. This MAb inhibits primary mixed lymphocyte culture (MLC) but not secondary MLC, cytotoxicity or proliferation induced by mitogens. It blocks binding of NK1-L16 stimulated T cells to L cells expressing CD50. This MAb is excellent for staining of formalin/paraffin tissues.

Type: Primary
Antigen: ICAM3
Clonality: Monoclonal
Clone: 101-1D2
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR