Imprimer

Anti-CCR5 Mouse Monoclonal Antibody (CF405S) [clone: 12D1.]

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BNC040156-500 BNC040156-100
BNC040156-500EA 560 EUR
BNC040156-500 BNC040156-100
Anti-CCR5 Mouse Monoclonal Antibody (CF405S) [clone: 12D1.]
Anticorps
Reacts with the N-terminal extracellular domain of CD195. The CC chemokine receptor 5 (CCR5) is a member of the CC-chemokine receptor family, and has the characteristic structure of a 7 transmembrane G protein-coupled receptor (GPCR). CCR5 regulates trafficking and effector functions of memory/effector Th1 cells, macrophages, NK cells, and immature dendritic cells. CCR5 and its ligands play an important role in viral pathogenesis. CCR5 represents the co-receptor for macrophage (M) and dual (T cell and M)-tropic immunodeficiency viruses. Together with the CD4 binding receptor, CCR5 plays a critical role in HIV entry into the target cells. Moreover, the CCR5 ligands macrophage inflammatory protein (MIP)-1 alpha, MIP-1 beta and RANTES act as endogenous inhibitors of HIV infection, making both CCR5 and its chemokine ligands attractive therapeutic targets for HIV infection. Recent studies have also highlighted the role of CCR5 in a variety of other human diseases, ranging from infectious and inflammatory diseases to cancer.

Type: Primary
Antigen: CCR5
Clonality: Monoclonal
Clone: 12D1.
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR