Imprimer
VWR® ARISTAR®, Antimony Standard Solutions for ICP-MS
Marques : VWR Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
456632HEA 212 EUR
456632H 457002K 85595.180
VWR® ARISTAR®, Antimony Standard Solutions for ICP-MS
Standards Cationic Standards Antimony Standards
  • Produced from high purity acids, water ASTM I 18 Megaohms
  • Solution assayed by titration
  • Final concentration verified against reference material from NIST
  • Total maximum uncertainty: 0,5% (typical)
  • Delivered with complete certificate of analysis
Matrix for 1000 and 10000 ppm contain a trace amount of tartaric acid.
Certifications: Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities