Imprimer
Human processed serum (HPS), hormone depleted
Fournisseur: SEQENS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
34000D-100ML 34000D-500ML
34000D-100MLEA 252 EUR
34000D-100ML 34000D-500ML
Human processed serum (HPS), hormone depleted
Sérums
SEQENS IVD manufactures Hormones Depleted Human Processed Serum.
  • Dry extract: 65 to 85 g/L
  • Total proteins: 55 to 75 g/L
  • Cortisol: <0,11 µg/dl
  • Total T3: <0,6 nmol/L
  • Total T4: <13 nmol/L
  • Free T3: <1,5 pmol/L
  • Free T4: <5,2 pmol/L
  • Filtered through a 0,2 µm membrane
  • Shelf life: 2 years
For this product, source material has been tested and found negative or non-reactive (FDA approved and/or CE marked test methods) for anti-HIV 1+2 and anti-HCV antibodies, HBs antigen, HIV 1 and HCV RNA and Syphilitic serology.
Complete donor traceability on file.
Attention: Only for research or manufacture of in vitro diagnostic reagents. Not for therapeutic use. This product of human origin should be considered as potentially infectious and handled accordingly.