Imprimer
Serum substitute, BaseSerum™
Fournisseur: SEQENS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
30001C-100ML 30001C-500ML 30001C-1L
30001C-100MLEA 155 EUR
30001C-100ML 30001C-500ML 30001C-1L
Serum substitute, BaseSerum™
Sérums
SEQENS IVD BaseSerum™ is a human serum substitute obtained from human plasma.
  • Total proteins: 60 to 80 g/L
  • Fibrinogen: Negative
  • Filtered through a 0,2 µm membrane
  • Shelf life: 2 years
It is electrolytes balanced to match physiological conditions. For this product, source material has been tested and found negative or non-reactive (FDA approved and/or CE marked test methods) for anti-HIV 1+2 and anti-HCV antibodies, HBs antigen, HIV 1 and HCV RNA and Syphilitic serology.
Complete donor traceability on file.
Attention: Only for research orEn savoir plus