Imprimer
Lymphocyte separation medium, reagent grade, LSM™, MP Biomedicals
Fournisseur: MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0850494X 0850494
ICNA0850494XEA 57.3 EUR
ICNA0850494X ICNA0850494
Lymphocyte separation medium, reagent grade, LSM™, MP Biomedicals
Milieux de séparation cellulaire
Lymphocyte Separation Medium (LSM™) is formulated for isolation of mononuclear cells from defibrinated or heparinised whole human blood. One-step centrifugation permits separation of mononuclear lymphocytes from erythrocytes, polynuclear lymphocytes and most platelets. It is a sterile-filtered solution of a sucrose polymer and diatrizoate salts.